

Iron(III) chloride, anhydrous, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7705-08-0
:
Formula:FeCl₃
MDL Number:MFCD00011005
:162.21 g/mol
Merck Index:13,04048
Boiling Pt:315 °C (1013 hPa)
Melting Pt:260 °C
Density:2.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:1773
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Dark green to black, cryst. powder (NK)
- Assay:Min. 98,5%
- Insoluble matter:Max. 0,2%
- Arsenic (As):Max. 0.001%
- Chromium (Cr):Max. 0,05%
- Copper (Cu):Max. 0,01%
- Manganese (Mn):Max. 0,2%
- Nickel (Ni):Max. 0,03%
- Lead (Pb):Max. 0.001%
- Zinc (Zn):Max. 0,04%
Specification Test Results
Frequently Bought Together