
Manganese(IV) oxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-13-9
:
Formula:MnO₂
MDL Number:MFCD00003463
:86.94 g/mol
Merck Index:14,5730
Melting Pt:535 °C (dec.)
Density:5.026
EINECS:215-202-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together