
Manganese(II) oxide 99% (76.0 - 78.0% Mn)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1344-43-0
:
Formula:MnO
MDL Number:MFCD00016226
:70.94 g/mol
Melting Pt:1840 °C
Density:5.0
EINECS:215-695-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together