Human Recombinant IGF-I (from E. coli)
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death. Mature human IGF-I is 100% homologous to bovine and porcine IGF-1 proteins.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: Human breast cancer cell line MCF-7, ED50 ≤1,5 ng/ml, Bioactivity ≥6,7×10⁵ units/mg
- Gene ID:P05019
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:mechano growth factor|IGF-IA|Somatomedin C
- Protein/peptide name:IGF-I
- Purity:≥95%
- Molecular weight:7,7 kDa
- Sequence:GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together