Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate, anhydrous ≥99.5% (by acidimetry), LiChropur™ for HPLC, Supelco®
Potassium dihydrogen phosphate, anhydrous ≥99.5% (by acidimetry), LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates
Potassium dihydrogen phosphate, anhydrous ≥99.5% (by acidimetry), LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidometric):
    ≥99,5%
  • Identity:
    Passes test
  • Ca (calcium):
    ≤0,50 ppm
  • Cu (copper):
    ≤0,005 ppm
  • Fe (iron):
    ≤0,10 ppm
  • Mg (magnesium):
    ≤0,50 ppm
  • Na (sodium):
    ≤20,0 ppm
  • HPLC gradient suitability test, chromophoric impurities based on 4'-hydroxyacetophenone:
    ≤10 ppm
  • Chromophore purity:
    ≥99,999%

Specification Test Results

Related Information

Frequently Bought Together