Order Entry
Austria
ContactUsLinkComponent
Human Recombinant IL-1RA (from E. coli)
Human Recombinant IL-1RA (from E. coli)
Certificates
undefined
Human Recombinant IL-1RA (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: Inhibition of human IL-1 alpha induced proliferation of D10,G4,1 cells, ED50 ≤50 ng/ml, bioactivity ≥2,0×10⁴ units/mg
  • Gene ID:
    P18510
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    IL-1F3|IL1RN
  • UniProtKB:
    P18510
  • Protein/peptide name:
    IL-1RA
  • Purity:
    ≥95%
  • Molecular weight:
    17,3 kDa
  • Sequence:
    MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together