About this item
Interleukin 10 (IL-10) is an anti-inflammatory cytokine produced by macrophages and type 2 T helper (Th2) cells. IL-10 inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFN-ƴ), tumor necrosis factor alpha (TNF-α), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. IL-10 also suppresses antigen presentation on antigen presenting cells, and enhances the survival, proliferation, and antibody production of B cells.
- High quality
- Low endotoxin
- Affordable
It inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFNƴ), tumor necrosis factor alpha (TNFα), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. Mouse IL-10 is not active on human cells
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P29456
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Cytokine synthesis inhibitory factor|CSIF
- UniProtKB:P29456
- Protein/peptide name:IL-10
- Purity:≥95%
- Molecular weight:18,8 kDa
- Sequence:MSKGHSIRGD NNCTHFPVSQ THMLRELRAA FSQVKTFFQK KDQLDNILLT DSLLQDFKGY LGCQALSEMI KFYLVEVMPQ AENHGPEIKE HLNSLGEKLK TLWIQLRRCH RFLPCENKSK AVEQVKNDFN KLQDKGVYKA MNEFDIFINC IEAYVTLKMK N
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT