Tungsten trioxide
Catalog # MSPP200008986
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pk:60 g
- Seq:3377
Specifications
About this item
:
:231.84 g/mol
Merck Index:12,09947
Melting Pt:1473 °C
Density:7.16 g/cm³ (20 °C)
EINECS:215-231-4
:GHS07
CAS Number:1314-35-8
Formula:O₃W
MDL Number:MFCD00011466