Order Entry
Austria
ContactUsLinkComponent
 
Aldehyde dehydrogenase (from Yeast)
 :  Merck
 
Aldehyde dehydrogenase (from Yeast)
 :  Merck
 

 

  • CAS No.:
    9028-88-0
  • Source:
    Yeast
  • Storage conditions:
    −20 °C
  • Enzyme name:
    Aldehyde dehydrogenase
  • pH:
    8,0

 

 

Frequently Bought Together