Order Entry
Austria
ContactUsLinkComponent
Poly(diallyldimethylammonium chloride) [pDADMAC] solution, M.W. <100000 35% (w/w) in water, very low molecular weight, Sigma-Aldrich®
Poly(diallyldimethylammonium chloride) [pDADMAC] solution, M.W. <100000 35% (w/w) in water, very low molecular weight, Sigma-Aldrich®
Supplier:  Merck
Certificates
Poly(diallyldimethylammonium chloride) [pDADMAC] solution, M.W. <100000 35% (w/w) in water, very low molecular weight, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Infrared spectrum:
    Conforms to structure
  • Viscosity (25 °C):
    100 - 200 cps
  • pH (c = neat):
    5.0 - 8.0
  • Total solids:
    34.0 - 36.0%
  • Concentration:
    35 wt. % in H₂O

Specification Test Results

Frequently Bought Together