ContactUsLinkComponent
Paraffin wax blended with synthetic polymers, Solidification point: 50-54°C, Surgipath® Paraplast Xtra
Paraffin wax blended with synthetic polymers, Solidification point: 50-54°C, Surgipath® Paraplast Xtra
  LEIC39603002
 :  
Paraffin wax blended with synthetic polymers, Solidification point: 50-54°C, Surgipath® Paraplast Xtra
  LEIC39603002
 :  
  • Pk:
    8 kg
  • Seq:
    4138
  • Pk Info:
    8× 1 kg
 :
 :1
 :8002-74-2
 :8012-95-1