Zinc chloride, Sigma-Aldrich®
Catalog # SIAL229997-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
About this item
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
:
:136.3 g/mol
Merck Index:13,10185
Boiling Pt:732 °C (1013 hPa)
Melting Pt:283 °C
Density:2.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
:GHS09
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295