Zinc chloride 97,0-100,5, granular USP, Multi-Compendial, J.T.Baker®
Catalog # 4326-05
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:2,5 kg
- Seq:4076
- Environmentally Preferable:
Specifications
Specification Test Results
- GMP Manufactured Product:Passes Test
- Meets U.S.P Requirements:Passes Test
- Meets E.P. Chemical Specifications:≤0.03%
- Meets J.P. Chemical Specifications:≤1.0%
- Meets B.P. Chemical Specifications:Passes Test
- Store at 2 to 25 °C:≤0.005%
- CAUTION: For Manufacturing, processing or repackaging:97.0 - 100.5%
- Bulk Pharmaceutical Chemical:≤10
- USP - Identification:Passes Test
- USP - Oxychloride:Passes Test
- USP - Sulfate (SO₄):≤0.03 %
- USP - Alkalies and Alkaline Earths:≤1.0 %
- USP - Ammonium Salts:Passes Test
- USP - Lead (Pb):≤0.005 %
- USP - Assay (ZnCl₂):97.0 - 100.5 %
- Endotoxin Concentration (EU/g):≤10
- EP/BP - Assay (ZnCl₂):95.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - pH:4.6 - 5.5
- EP/BP - Oxychloride:Passes Test
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Aluminum, Calcium, Heavy Metals, Iron and Magnesium:Passes Test
- EP/BP - Ammonium (NH₄):≤400 ppm
- JP - Assay (ZnCl₂):97.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤0.010 %
- JP - Ammonium (NH₄):Passes Test
- JP - Heavy Metals (as Pb):≤50 ppm
- JP - Alkalies and Alkaline Earths:≤1.00 %
- JP - Arsenic (As):≤5 ppm
- JP - Oxychloride:Passes Test
Specification Test Results
About this item
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
:
:136.3 g/mol
EINECS:231-592-0
UN:2331
:GHS05
:GHS07
:GHS09
:GHS08
Storage Temperature:Ambient
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295
Boiling Pt:732
Melting Pt:283
Density:2.91 g/cm³ (20 °C)
ADR:8 III
Merck Index:13,10185