Mercury(II) chloride, fine crystals, EMPLURA®, Supelco®
Catalog # 1.04417.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (complexometric; calculated on dried substance):99.5 - 100.5 %
- Identity:passes test
- Appearance of solution:passes test
- Acidity or alkalinity:passes test
- Mercury(I)-chloride:passes test
- Loss on drying:≤ 1.0 %
- Density:5.44 g/cm3 (20 °C)
- pH:3.2 (15 g/l, H2O)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride
:
:271.5 g/mol
EINECS:231-299-8
UN:1624
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:7487-94-7
Formula:HgCl₂
MDL Number:MFCD00011041
Boiling Pt:302
Melting Pt:273
Density:5.44 g/cm³ (20 °C)
ADR:6.1 II
Merck Index:13,05901
:M-Clarity™ quality level = MQ200