



Mercury(II) chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7487-94-7
:
Formula:HgCl₂
MDL Number:MFCD00011041
:271.5 g/mol
Merck Index:14,5876
Boiling Pt:302 °C
Melting Pt:280…282 °C
Density:5.4
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together