
Silver sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (1013 hPa)
Melting Pt:655 °C
Density:5.45 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-653-7
:GHS05
Storage Temperature:Ambient
Frequently Bought Together