
Silver sulphate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (1013 hPa)
Melting Pt:655 °C
Density:5.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-653-7
UN:3077
ADR:9,III
:GHS05
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
Frequently Bought Together