Order Entry
Austria
ContactUsLinkComponent
Silver sulphate ACS analytical reagent, Supelco®
Corrosive
Environment
Silver sulphate ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Environment
Silver sulphate ACS analytical reagent, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    8 g/l (25 °C)
  • Melting Point:
    655 °C
  • Molar Mass:
    311.8 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    5.45 g/cm3 (20 °C)
  • pH:
    5 - 6 (5 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together