
Silver sulphate ≥99.5%, Puriss. p.a. ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (1013 hPa)
Melting Pt:655 °C
Density:5,45 g/cm³ (20 °C)
EINECS:233-653-7
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.5 %
- Water insoluble matter and silver chloride:Max. 0.02 %
- Substances not precipitated by HCl:Max. 0.03 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 50 ppm
- Cobalt (Co):Max. 50 ppm
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 100 ppm
- Sodium (Na):Max. 150 ppm
- Nickel (Ni):Max. 0.001 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.0005 %
- Chloride (Cl):Max. 0.001 %
- Nitrate (NO₃):Max. 0.001 %
Specification Test Results
Frequently Bought Together