

Silver sulphate ≥98% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:14,8529
Boiling Pt:1085 °C (dec.)
Melting Pt:657 °C
Density:5.45
Storage Temperature:Ambient
EINECS:233-653-7
:GHS05
:GHS09
Related Information
Frequently Bought Together