Copper(II) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
Boiling Pt:2000
Melting Pt:1326
Density:4.9 g/cm³ (20 °C)
Molecular Weight:79.55 g/mol
EINECS:215-269-1
UN:3077
ADR:9 III
Merck Index:13,02674
:M-Clarity™ quality level = MQ100
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together