Copper(II) oxide ACS analytical reagent, Supelco®
: Merck
: Cupric oxide, Copper oxide, Copper monoxide granulated
:1317-38-0
:
:CuO
:MFCD00010979
:2000
:1326
:4.9 g/cm³ (20 °C)
:79.55 g/mol
:215-269-1
:3077
:9 III
:13,02674
:M-Clarity™ quality level = MQ300
:GHS09
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) insoluble
- Melting Point:1326 °C (decomposition)
- Molar Mass:79.55 g/mol
- Bulk Density:500 kg/m3
- Density:6.48 g/cm3 (25 °C)
- pH:7 (50 g/l, H2O, 20 °C) (slurry)
Frequently Bought Together