Copper(II) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
Boiling Pt:2000
Melting Pt:1326
Density:6.3
Molecular Weight:79.55 g/mol
Storage Temperature:Ambient
EINECS:215-269-1
ADR:9 III
Merck Index:14,2646
UN:3077
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together