

Copper(I) oxide, red ≥97%, TECHNICAL
Catalog # 23135.295
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 97 %
Specification Test Results
About this item
:
:143.09 g/mol
Merck Index:13,02694
Boiling Pt:1800 °C (1013 hPa)
Melting Pt:1232 °C
Density:5.7…6.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-270-7
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:1317-39-1
Formula:Cu₂O
MDL Number:MFCD00010974