
Copper(I) iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7681-65-4
:1335-23-5
Formula:CuI
MDL Number:MFCD00010978
:190.45 g/mol
Merck Index:13,02692
Boiling Pt:1290 °C (1013 hPa)
Melting Pt:602 °C
Density:5.62 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:231-674-6
:GHS09
Storage Temperature:Ambient
Frequently Bought Together