Order Entry
Austria
ContactUsLinkComponent
Copper(I) cyanide 99%, extra pure
Copper(I) cyanide 99%, extra pure
Certificates
undefined
Copper(I) cyanide 99%, extra pure

Specification Test Results

  • Appearance:
    White to beige to green Powder
  • Assay:
    ≥98.5 % (ex Cu)
  • Arsenic (As):
    ≤10 ppm
  • Iron (Fe):
    ≤100 ppm
  • Lead (Pb):
    ≤20 ppm
  • Tin (Sn):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together