


Copper(II) chloride for synthesis, Sigma-Aldrich®
: Merck
:7447-39-4
:
:Cl₂Cu
:MFCD00010972
:134.45 g/mol
:13,02660
:1365.85 °C (1013 hPa)
:628 °C
:3.054 g/cm³ (20 °C)
:Ambient
:2802
:8,III
:M-Clarity™ quality level = MQ200
:231-210-2
:GHS05
:GHS07
:GHS09
- Solubility:620 g/l (20 °C)
- Melting Point:498 °C decomposes
- Molar Mass:134.45 g/mol
- Density:3.39 g/cm3 (25 °C)
- pH:3.5 (50 g/l, H2O, 20 °C)
- Synonyms:Copper dichloride
- Hill Formula:Cl2Cu
- Chemical Formula:CuCl2
Frequently Bought Together