Orders LinkContactUsLinkComponent
Manual crimping tools, advanced ergonomic version
Manual crimping tools, advanced ergonomic version
Supplier:   MACHEREY-NAGEL
Manual crimping tools, advanced ergonomic version
Supplier:  MACHEREY-NAGEL
Certificates
Manual crimping tools, advanced ergonomic version
Manual crimping tools, advanced ergonomic version
5 Options Available Below
Product Details & Documents
These manual crimping tools with ergonomically designed handles are more lightweighted than complete steel crimpers.
Ergonomic crimpers:
• Adjustable knob on the crimping head that is easily accessible and visible
• Activated by bottom handle motion only which allows a steadier and safer hold of the tool during crimping
• Crimping head design and alignment makes better vertical clearance over the vial
Ergonomic decappers:
• Allow safe removal of caps
• No adjustment required
Product Family Options
Manual crimpers, ergonomic
Product Information
  • Availability
    Price
  • MANA735208
    Ergonomic crimper for 8 mm aluminium crimp caps
    Ergonomic crimper for 8 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 266,00 €
    266,00 €
    1 * 1 ST
       266,00 €
  • MANA735211
    Ergonomic crimper for 11 mm aluminium crimp caps
    Ergonomic crimper for 11 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
  • MANA735220
    Ergonomic crimper for 20 mm aluminium crimp caps
    Ergonomic crimper for 20 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
Manual decappers, ergonomic
Product Information
  • Availability
    Price
  • MANA735311
    Ergonomic decapper for 11 mm aluminium crimp caps
    Ergonomic decapper for 11 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
  • MANA735320
    Ergonomic decapper for 20 mm aluminium crimp caps
    Ergonomic decapper for 20 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
Manual decappers, ergonomic
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Ergonomic decapper for 11 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Ergonomic decapper for 20 mm aluminium crimp caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €