Microscale tissue grinder kit, KIMBLE®
Artikelnr. SCERSP884900-0000
Lieferant: DWK Life Sciences
Spezifikationen
- Description:Microscale tissue grinder kit
- Pk:1
Spezifikationen
Über diesen Artikel
The kit contains a variety of the finest microscale tissue grinders suitable for use on tough connective tissue and soft tissue such as brain or liver.
- Working with micro quantities reduces heat build-up and yields greater enzyme activity
- Samples can be handled with minimal loss; transfers can often be eliminated
- HDPE case with die cut foam inserts that protect grinders in transit and in storage
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Kit includes ground glass, smooth glass and PTFE grinders.