Order Entry
Austria
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.9% ACS, Sigma-Aldrich®
Dimethyl sulphoxide ≥99.9% ACS, Sigma-Aldrich®
Catalog # SIAL472301-100ML
Supplier:  Merck
CAS Number:  
Dimethyl sulphoxide ≥99.9% ACS, Sigma-Aldrich®
Catalog # SIAL472301-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089