Order Entry
Austria
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.5% stationary phase for GC
Dimethyl sulphoxide ≥99.5% stationary phase for GC
Catalog # 23487.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.5% stationary phase for GC
Catalog # 23487.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089