Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay:Min. 99,0 %
- IR Spectrum:Passes test
- Density (20/4):1,099 - 1,101
- Refractive index (20°C):1,478 - 1,479
- Evaporation residue :Max. 50 ppm
- Water :Max. 0.05 %
- Cu (Copper) :Max. 5 ppm
- Fe (Iron) :Max. 5 ppm
- Pb (Lead) :Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
Packaging Information
spec_VWRC23486.297 spec_VWRC23486.322 spec_VWRC23486.460
Product specification
Frequently Bought Together