Order Entry
Austria
ContactUsLinkComponent
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay:
    min. 99.0 %
  • Color (APHA):
    max. 10
  • Density (g/ml) at 25°C:
    min. 1.095
  • Freezing Point:
    18.0 - 19.0 °C
  • Residue after Evaporation:
    max. 0.01 %
  • Water (H2O):
    max. 0.2 %

Specification Test Results

Related Information

Frequently Bought Together