Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Pyrogen test:passes test
- Assay (GC):min. 99.5 %
- Free acid:max. 0.001 %
- Non-volatile matter:max. 0.001 %
- Total P:max. 0.00001 %
- Total Si:max. 0.00002 %
- Water (K.F.):max. 0.1 %
- Ca:max. 0.0001 %
- Cu:max. 0.00005 %
- Fe:max. 0.0001 %
- K:max. 0.00005 %
- MG:max. 0.00005 %
- NA:max. 0.0002 %
- PB:max. 0.00002 %
- Zn:max. 0.00002 %
Specification Test Results
Frequently Bought Together