Order Entry
Austria
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.9% for HPLC
Catalog # 42780.AK
   Safer Human and Environmental Health
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.9% for HPLC
Catalog # 42780.AK
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

About this item

CAS Number:67-68-5
 :
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
 :78.14 g/mol
Merck Index:14,3259
Boiling Pt:189 °C
Melting Pt:18.4 °C
Density:1.101
 :Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:87 °C (189 °F)
Storage Temperature:Ambient
EINECS:200-664-3