0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Dimethyl sulphoxide
Dimethyl sulphoxide ≥99.9% for spectrophotometry
Dimethyl sulphoxide ≥99.9% for spectrophotometry
:
Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% for spectrophotometry
:
Thermo Fisher Scientific
:
67-68-5
:
:
(CH₃)₂SO
:
MFCD00002089
:
78.14 g/mol
:
14,3259
:
189 °C
:
18.4 °C
:
1.101
:
87 °C (189 °F)
:
Ambient
:
200-664-3
msds_ALFA032434
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
N,N-Dimethylformamide, SupraSolv® for headspace gas chromatography, Supelco®
: Merck
D-(-)-Mannitol 97.0-102.0% USP, Ph. Eur.
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Sulphuric acid 96% ≥96%, Suprapur® for trace analysis, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
: Merck