Dimethyl sulphoxide 99.5+% for analysis
:67-68-5
:
:(CH₃)₂SO
:MFCD00002089
:78.14 g/mol
:13,03285
:189 °C (760 mmHg)
:18.4 °C
:1.1 g/cm³
:87 °C
:Ambient
:200-664-3
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.50 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Substance darkened by H2SO4:Passes test
- Iron (Fe):≤1 ppm
Frequently Bought Together