Dimethyl sulphoxide 99.8+% for molecular biology
Catalog # ACRO327182500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.80 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.001 %
- Dnase,Rnase,Protease activity:none detected
Specification Test Results
About this item
:
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.4 °C
Density:1.1 g/cm³
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089