ContactUsLinkComponent
Dimethyl sulphoxide 99.8+%, extra pure
Dimethyl sulphoxide 99.8+%, extra pure
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide 99.8+%, extra pure
   Sustainable Options Available
Certificates
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 %
  • GC:
    ≥99.80 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.003 %
  • Acidity:
    ≤0.04 mg KOH/g