
Ammonium sulphate 99.0-100.5%, granular, BAKER ANALYZED® FCC, ACS for biochemistry, for chromatography endotoxin tested, J.T.Baker®
Catalog # 0793-08
Supplier: Avantor
CAS Number:
Specifications
- Pack type:PE drum, open head
- Pk:325 lb
- Seq:7239
Specifications
Specification Test Results
- Food GMP Manufactured Product:99.0 - 100.5%
- Meets ACS Reagent Chemical Requirements:≤0.005%
- Meets F.C.C. Requirements:≤0.005%
- CAUTION: For Manufacturing, processing or repackaging:5.0 - 6.0
- Bulk Food Chemical:≤0.001%
- ACS - Assay ((NH₄)₂SO₄):99.0 - 100.5 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Residue on Ignition (ACS):≤0.005 %
- ACS - pH of 5% Solution at 25°C:5.0 - 6.0
- ACS - Nitrate (NO₃):≤0.001 %
- Chloride (Cl):≤3 ppm
- Phosphate (PO₄):≤3 ppm
- Heavy Metals (as Pb):≤3 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- FCC - Assay ((NH₄)₂SO₄):99.0 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Residue on Ignition (FCC):≤0.25 %
- FCC - Lead (Pb):≤3 mg/kg
- FCC - Selenium (Se):≤0.003 %
- Endotoxin Concentration (2.5 EU/g max.):Passes Test
Specification Test Results
About this item
:
:132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
EINECS:231-984-1
REACH:01-2119455044-46
:GHS08
Storage Temperature:Ambient
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391