Order Entry
Austria
ContactUsLinkComponent
Ammonium peroxodisulphate (APS)
Ammonium peroxodisulphate (APS)
Catalog # BMOL50404-1
CAS Number:  
undefined
Ammonium peroxodisulphate (APS)
Catalog # BMOL50404-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate
 :
 :228.2 g/mol
EINECS:231-786-5
UN:1444
 :GHS03
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:7727-54-0
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
Melting Pt:120
Density:1.98 g/cm³ (20 °C)
ADR:5.1 III
Merck Index:13,00545