Order Entry
Austria
ContactUsLinkComponent
 
Ammonium dihydrogen phosphate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
Catalog # 17842-250G
CAS Number:  
Ammonium dihydrogen phosphate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
Catalog # 17842-250G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance of the solution (c=10%):
    complying
  • Assay:
    99.0 - 101.0 %
  • pH (10 %, 20°C):
    4.0 - 5.0
  • Absorbance at 210 nm:
    Max. 0.05
  • Absorbance at 220 nm:
    Max. 0.04
  • Absorbance at 230 nm:
    Max. 0.03
  • Absorbance at 240 nm:
    Max. 0.02
  • Absorbance 500 nm:
    Max. 0.02

Specification Test Results

About this item

 :
Merck Index:13,00547
Boiling Pt:197 °C (Decomposition)
Melting Pt:190 °C
Density:1.803 g/cm³ (20 °C)
 :115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Boiling Pt:197 °C (dec.)