Order Entry
Austria
ContactUsLinkComponent
Ammonium dihydrogen phosphate, Suprapur® for trace analysis, Supelco®
Ammonium dihydrogen phosphate, Suprapur® for trace analysis, Supelco®
Catalog # 1.01440.0050
Supplier:  Merck
CAS Number:  
Ammonium dihydrogen phosphate, Suprapur® for trace analysis, Supelco®
Catalog # 1.01440.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    368 g/L (20 °C)
  • Melting Point:
    190 °C
  • Molar Mass:
    115.02 g/mol
  • Bulk Density:
    800 - 1100 kg/m³
  • Vapor Pressure:
    0.066 hPa (125 °C)
  • Density:
    1.80 g/cm³ (20 °C)
  • pH:
    3.8 - 4.4 (50 g/L, H₂O, 20 °C)

Specification Test Results

About this item

 :
Merck Index:13,00547
Melting Pt:190 °C
Density:1.8 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396