Order Entry
Austria
ContactUsLinkComponent
Diammonium hydrogen orthophosphate ≥98%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Diammonium hydrogen orthophosphate ≥98%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Diammonium hydrogen orthophosphate ≥98%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • pH (20°C):
    About 8
  • pH (25°C; 5 %):
    7.7 - 8.1
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 0.001 %
  • SO4 (Sulphate):
    Max. 0.01 %
  • NO3 (Nitrate):
    Max. 0.003 %
  • Ca (Calcium):
    Max. 10 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 0.005 %
  • Mg (Magnesium):
    Max. 5 ppm
  • Na (Sodium):
    Max. 0.005 %
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together