Order Entry
Austria
ContactUsLinkComponent
Diammonium hydrogen orthophosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Diammonium hydrogen orthophosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Diammonium hydrogen orthophosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    690 g/l (20 °C)
  • Molar Mass:
    132.05 g/mol
  • Bulk Density:
    800 - 1000 kg/m3
  • Density:
    1.619 g/cm3 (20 °C)
  • pH:
    7.6 - 8.2 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together