
Ammonium carbonate Ph. Franç.
83516.290
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay (NH3):30.0 - 34.0 %
- Appearance:White crystalline powder
- Identification A:Passes test
- Identification B:Passes test
- Identification C:Passes test
- Appearance of solution:Passes test
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Sulphated ash:Max. 0.05 %
:
:96.09 g/mol
:13,00508
:58 °C
:1.5 g/cm³ (20 °C)
:208-058-0
:GHS07
:Ambient
:10361-29-2
:(NH₄)₂CO₃
:MFCD00010890