

Ammonium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:233-786-0
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (acidimetric, NH₃):≥ 30.0 %
- Identity:passes test
- Description:white crystals
- Insoluble matter:≤ 0.005 %
- Chloride (Cl):≤ 0.0005 %
- Sulfur compounds (as SO₄):≤ 0.002 %
- Heavy metals (as Pb):≤ 0.0005 %
- Fe (Iron):≤ 0.0005 %
- Non-volatile matter:≤ 0.01 %
- Conforms ACS, Reag. Ph. Eur.:passes test
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.59504.0250_TDS_EN MERC1.59504.1000_TDS_EN ProductPage_159504
Product specification
Frequently Bought Together