
Ammonium carbonate, puriss. NF, Ph. Franç., FCC, extra pure, Sigma-Aldrich®
Catalog # SIAL11204-25KG-H
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:8000-73-5
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890