
Ammonium carbonate, Ultrapure
Catalog # 0305-1KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (NH3):>30%
- Chloride:<0.0005%
- Heavy Metals (as Pb):<0.0005%
- Insolubles:<0.005%
- Iron:<0.0005%
- Non-volatile Matter:<0.01%
- Sulphur Compounds:<0.002%
Specification Test Results
About this item
:
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890