Ammonium carbonate for analysis
:506-87-6
:10361-29-2
:(NH₄)₂CO₃
:MFCD00010890
:96.09 g/mol
:13,00508
:58 °C (dec.)
:1,5 g/cm³ (20 °C)
:208-058-0
:GHS07
:Ambient
- Appearance:White Powder or crystals and/or chunks
- Non-volatile matter:≤0.01 %
- Titration with HCl:≥30.0 % (NH3)
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
Frequently Bought Together